EMAIL THIS PAGE TO A FRIEND
442379 Supelco

3,5-Dimethylphenol

analytical standard

Synonym: 5-Hydroxy-m-xylene, sym.-m-Xylenol

  • CAS Number 108-68-9

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein Registry Number 774117

  •  EC Number 203-606-5

  •  MDL number MFCD00002307

  •  PubChem Substance ID 24867822

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
InChI Key   TUAMRELNJMMDMT-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   222 °C(lit.)
mp   61-64 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
2
RTECS 
ZE6475000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?