EMAIL THIS PAGE TO A FRIEND
442394 Supelco

3-Nitrotoluene

analytical standard

Synonym: 1-Methyl-3-nitrobenzene

  • CAS Number 99-08-1

  • Linear Formula CH3C6H4NO2

  • Molecular Weight 137.14

  •  Beilstein Registry Number 1906910

  •  EC Number 202-728-6

  •  MDL number MFCD00007265

  •  PubChem Substance ID 24867828

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aromatics, Chemical Class,
grade   analytical standard
vapor density   4.73 (vs air)
vapor pressure   1 mmHg ( 50.2 °C)
InChI Key   QZYHIOPPLUPUJF-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.541(lit.)
bp   230-231 °C(lit.)
mp   14-16 °C(lit.)
density   1.157 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1664 6.1 / PGII
WGK Germany 
3
RTECS 
XT2975000
Flash Point(F) 
215.6 °F
Flash Point(C) 
102 °C
Protocols & Articles

Articles

EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography

GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitroamines, Mix B on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitroamines, Mix B on Ascentis® C18
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?