EMAIL THIS PAGE TO A FRIEND
442405 Supelco

4-Bromophenol

analytical standard

  • CAS Number 106-41-2

  • Linear Formula BrC6H4OH

  • Molecular Weight 173.01

  •  Beilstein Registry Number 1680024

  •  EC Number 203-394-4

  •  MDL number MFCD00002313

  •  PubChem Substance ID 24867834

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
InChI Key   GZFGOTFRPZRKDS-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   235-236 °C(lit.)
mp   61-64 °C(lit.)
format   neat
storage temp.   room temp
Gene Information   human ... ALOX12(239), ALOX15(246)

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
SJ7960000
Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?