EMAIL THIS PAGE TO A FRIEND
442445-U Supelco

Allyl alcohol

analytical standard

Synonym: 2-Propen-1-ol

Purchase

Properties

Related Categories A-D, All Herbicide Standards, Analytical Standards, Analytical/Chromatography, Herbicides,
grade   analytical standard
vapor density   2 (vs air)
vapor pressure   23.8 mmHg ( 25 °C)
InChI Key   XXROGKLTLUQVRX-UHFFFAOYSA-N
form   neat
autoignition temp.   712 °F
expl. lim.   18 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.412(lit.)
bp   96-98 °C(lit.)
mp   −129 °C(lit.)
density   0.854 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Used to induce a mouse model of liver damage that has been used to study the mechanisms of hepatotoxicity and hepatic stem cell-mediated repair.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1098 - class 6.1 - PG 1 - Allyl alcohol
WGK Germany 
2
RTECS 
BA5075000
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles

Articles

GC Analysis of Alcoholic Spirit on Equity®-1 Coupled to SUPELCOWAX 10 (Column Set 2)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Alcoholic Spirit on Equity®-1 Coupled to SUPELCOWAX 10 (Column Set 2)
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?