EMAIL THIS PAGE TO A FRIEND
442446 Supelco

3-Chloro-1-propene

analytical standard

Synonym: 3-Chloro-1-propene, Allyl chloride, Chlorallylene

  • CAS Number 107-05-1

  • Linear Formula CH2=CHCH2Cl

  • Molecular Weight 76.52

  •  Beilstein Registry Number 635704

  •  EC Number 203-457-6

  •  MDL number MFCD00000984

  •  PubChem Substance ID 24867856

Purchase

Properties

Related Categories A-C, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor density   2.6 (vs air)
vapor pressure   20.58 psi ( 55 °C)
  5.71 psi ( 20 °C)
InChI Key   OSDWBNJEKMUWAV-UHFFFAOYSA-N
form   neat
expl. lim.   11.2 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.414(lit.)
bp   44-46 °C(lit.)
mp   −130 °C(lit.)
density   0.939 g/mL at 25 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Target organs 
Kidney, Liver, Nervous system
RIDADR 
UN1100 - class 3 - PG 1 - Allyl chloride
WGK Germany 
2
RTECS 
UC7350000
Flash Point(F) 
-25.6 °F
Flash Point(C) 
-32 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?