• USA Home
  • 442496-U - Bromochloroacetic acid

EMAIL THIS PAGE TO A FRIEND
442496-U Supelco

Bromochloroacetic acid

analytical standard

  • CAS Number 5589-96-8

  • Linear Formula BrClCHCOOH

  • Molecular Weight 173.39

  •  Beilstein Registry Number 1720556

  •  MDL number MFCD00143872

  •  PubChem Substance ID 329756470

Purchase

Properties

Related Categories A-C, Additional Standards, Analytical Standards, Analytical/Chromatography, Bromo,
grade   analytical standard
InChI Key   GEHJBWKLJVFKPS-UHFFFAOYSA-N
form   neat
packaging   ampule of 50 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.51(lit.)
bp   210-212 °C/767 mmHg(lit.)
mp   27.5 °C(lit.)
density   1.985 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
RTECS 
AF5957282
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?