EMAIL THIS PAGE TO A FRIEND
442538 Supelco

Decafluorobiphenyl

analytical standard

  • CAS Number 434-90-2

  • Linear Formula C6F5C6F5

  • Molecular Weight 334.11

  •  Beilstein Registry Number 1916978

  •  EC Number 207-107-3

  •  MDL number MFCD00000292

  •  PubChem Substance ID 24867895

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
InChI Key   ONUFSRWQCKNVSL-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   206 °C(lit.)
mp   68-70 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3152PSN1 9 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?