EMAIL THIS PAGE TO A FRIEND
442550-U Supelco

Dibenzothiophene

analytical standard

Synonym: DBT, Diphenylene sulfide

  • CAS Number 132-65-0

  • Empirical Formula (Hill Notation) C12H8S

  • Molecular Weight 184.26

  •  Beilstein Registry Number 121101

  •  EC Number 205-072-9

  •  MDL number MFCD00004969

  •  PubChem Substance ID 329756475

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, D, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   analytical standard
InChI Key   IYYZUPMFVPLQIF-UHFFFAOYSA-N
form   neat
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   332-333 °C(lit.)
mp   97-100 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
HQ3490550
Protocols & Articles

Articles

GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents GC Analysis of Polynuclear Aromatic Hydrocarbons (PAHs) in Salmon on SPB®-608 (20 m x 0.18 mm I.D., 0.18 μm) after QuEChERS Cleanup using Supel™ QuE Z-Sep, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?