EMAIL THIS PAGE TO A FRIEND
442665 Supelco

1-Butanol

analytical standard

Synonym: n-Butanol, Butyl alcohol

  • CAS Number 71-36-3

  • Linear Formula CH3(CH2)3OH

  • Molecular Weight 74.12

  •  Beilstein Registry Number 969148

  •  EC Number 200-751-6

  •  MDL number MFCD00002964

  •  PubChem Substance ID 24867944

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alcohols, Aliphatic,
grade   analytical standard
vapor density   2.55 (vs air)
vapor pressure   5 (4 mmHg) at 20 °C hPa
InChI Key   LRHPLDYGYMQRHN-UHFFFAOYSA-N
form   neat
autoignition temp.   649 °F
expl. lim.   11.2 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   colorless
refractive index   n20/D 1.399(lit.)
bp   116-118 °C(lit.)
mp   −90 °C(lit.)
density   0.81 g/mL at 25 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1120 - class 3 - PG 3 - Butanols
WGK Germany 
1
RTECS 
EO1400000
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C
Protocols & Articles

Articles

GC Analysis of Alcoholic Spirit on Equity®-1 Coupled to SUPELCOWAX 10 (Column Set 2)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Alcoholic Spirit on Equity®-1 Coupled to SUPELCOWAX 10 (Column Set 2)
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Alcohols on PAG

From our library of Articles, Sigma-Aldrich presents GC Analysis of Alcohols on PAG
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Class 3 Residual Solvents on OVI-G43

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on OVI-G43
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on SUPELCOWAX® 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on SUPELCOWAX® 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography

GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Class 3 Residual Solvents on VOCOL® after SPME using 100 μm PDMS Fiber
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Pharmaceutical, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Industrial Solvents on Equity®-1701

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on Equity®-1701
Keywords: Chromatography, Flame ionization detector, Gas chromatography

GC Analysis of Industrial Solvents on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on Equity®-1
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Industrial Solvents on SUPELCOWAX 10

From our library of Articles, Sigma-Aldrich presents GC Analysis of Industrial Solvents on SUPELCOWAX 10
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of a 56-Component Industrial Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column

From our library of Articles, Sigma-Aldrich presents GC Analysis of a 56-Component Industrial Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Keywords: Chromatography, Flame ionization detector, Gas chromatography

GC Analysis of an Indoor Air Standard on Equity®-1

From our library of Articles, Sigma-Aldrich presents GC Analysis of an Indoor Air Standard on Equity®-1
Keywords: Air Monitoring, Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?