• USA Home
  • 44671-U - EPA 625/8270C SV Surrogate Spike Mix with Indicator

EMAIL THIS PAGE TO A FRIEND
44671-U Supelco

EPA 625/8270C SV Surrogate Spike Mix with Indicator

analytical standard

Purchase

Properties

Related Categories 600 Series Wastewater Methods, 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type surrogate
packaging   ampule of 25 mL
concentration   in methanol (varied)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

2-Fluorobiphenyl 100 μg/mL SDS    
2-Fluorophenol 200 μg/mL SDS    
Nitrobenzene-d5 100 μg/mL SDS    
Phenol-d5 200 μg/mL SDS    
p-Terphenyl-d14 100 μg/mL SDS    
2,4,6-Tribromophenol SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?