• USA Home
  • 44981-U - SPME Performance Evaluation Mix

EMAIL THIS PAGE TO A FRIEND
44981-U Supelco

SPME Performance Evaluation Mix

1 μg/mL each component in deionized water: methanol (99:1), analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Column & Instrument Performance Test Mixes, GC Column Test Mixes More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 3 × 1 mL
concentration   1 μg/mL each component in deionized water: methanol (99:1)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Nitrobenzene SDS    
2-Nitrotoluene SDS    
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?