46842-U Supelco

EPA 8260 Surrogate Standards Mix 2

250 μg/mL each component in methanol, analytical standard

DOWNLOAD MSDS (PDF)
  •  EC Number 200-659-6

Properties

Related Categories 8000 Series Solidwaste Methods, Analytical Standards, Analytical/Chromatography, Chromatography, EPA,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type surrogate
packaging   ampule of 5 mL
concentration   250 μg/mL each component in methanol
suitability   suitable for 8260 per US EPA

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Analyte

Description

Product #

Add to Cart

4-Bromofluorobenzene    
Dibromofluoromethane    
1,2-Dichloroethane-d4    
Toluene-d8    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?