46863-U Supelco

Pesticide Mix B-2

in hexane: toluene (98.5:1.5) (varied), analytical standard

DOWNLOAD MSDS (PDF)

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Chromatography, Environmental Standards, Solid Waste More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   in hexane: toluene (98.5:1.5) (varied)
storage temp.   2-8°C

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Analyte

Description

Product #

Add to Cart

Aldrin 5 μg/mL    
β-BHC    
δ-BHC    
α-Chlordane 5 μg/mL    
γ-Chlordane 5 μg/mL    
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene 10 μg/mL    
Decachlorobiphenyl 10 μg/mL    
β-Endosulfan 10 μg/mL    
Endosulfan sulfate 10 μg/mL    
Endrin aldehyde 10 μg/mL    
Endrin ketone 10 μg/mL    
Heptachlor exo-epoxide 5 μg/mL    
2,4,5,6-Tetrachloro-m-xylene 5 μg/mL    

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?