CAS Number 303-47-9
Beilstein Registry Number 8169012
MDL number MFCD00078079
PubChem Substance ID 24870560
| Related Categories | All alphabetically sorted, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Biotoxins, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 50 μg/mL in benzene: acetic acid (99:1) |
| storage temp. | −20°C |
Customers Also Viewed
analytical standard, in methanol (varied)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?