Skip to Content
MilliporeSigma

47281

1,3,5-Trimethylbenzene

Synonym(s):

Mesitylene, 1,3,5-Trimethylbenzene

Sign In to View Organizational & Contract Pricing

About This Item

Linear Formula:
C6H3(CH3)3
CAS Number:
Molecular Weight:
120.19
Beilstein/REAXYS Number:
906806
EC Number:
MDL number:
UNSPSC Code:
41116107

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor density

4.1 (vs air)

vapor pressure

14 mmHg ( 55 °C)
2.49 mmHg ( 25 °C)

autoignition temp.

1022 °F

expl. lim.

0.88-6.1 %, 100 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.499 (lit.)

bp

163-166 °C (lit.)

mp

−45 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

application(s)

environmental

SMILES string

Cc1cc(C)cc(C)c1

InChI

1S/C9H12/c1-7-4-8(2)6-9(3)5-7/h4-6H,1-3H3

InChI key

AUHZEENZYGFFBQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalit Rechavi et al.
Journal of the American Chemical Society, 126(25), 7738-7739 (2004-06-24)
Isotope effects for the encapsulation of deuterated versus nondeuterated guests are determined. This system involves the use of social isomers, and the origin of the isotope effect is localized to methyl groups interacting with aromatic rings.
Toshifumi Dohi et al.
Chemical communications (Cambridge, England), (40)(40), 4152-4154 (2007-10-11)
We have found that the use of fluoroalcohol media greatly enhanced the efficiency and scope of the direct dehydrative condensation of arenes and hypervalent iodine(III) compounds; the present clean method has a broad range of applicability as well as unique
Claudio Evangelisti et al.
Journal of nanoscience and nanotechnology, 8(4), 2096-2101 (2008-06-25)
Pt nanoclusters have been generated by reaction of Pt vapour and mesitylene vapour and the role of the mesitylene/platinum ratio and the Pt particle size has been evaluated, quenching the resulting mesitylene solvated Pt atoms with 1,3-divinyltetramethyldisiloxane (DVS) as additional
Jacqueline M Veauthier et al.
Inorganic chemistry, 43(4), 1220-1228 (2004-02-18)
New bimetallic mu-oxo diferric complexes of several previously reported calix[4]pyrrole Schiff base macrocycles are described. The synthesis of a new member of this class of macrocycles is also reported; it was prepared via an acid-catalyzed condensation between 1,9-bisformyl-5,5-dipropyldipyrromethane and o-phenylenediamine.
Sanyog Sharma et al.
Organic & biomolecular chemistry, 11(4), 654-661 (2012-12-12)
Two mesitylene based probes, having catechol/phenol units in conjunction to the Schiff base have been synthesized. The probe with a catechol unit can be used to sense and discriminate between F(-) and CN(-) through different colorimetric and fluorimetric responses using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service