Synonym:
Butyraldehyde-
CAS Number 1527-98-6
EC Number 200-835-2
| Related Categories | Air Monitoring Standards, Aldehyde/DNPH, Aldehydes / DNPH / DAIH, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| feature | standard type calibration |
| packaging | ampule of 1 mL |
| concentration | 1000 μg/mL in acetonitrile (as aldehyde equivalent) |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?