EMAIL THIS PAGE TO A FRIEND
47505-U Supelco

BTEX/MTBE Mix, HC

2000 μg/mL each component in methanol, analytical standard

Purchase

Properties

grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
storage temp.   2-8°C

Description

Other Notes

This product is now available as a certified reference solution. Please reference CRM47505

Price and Availability


View Details on the New CRM Product

Analyte

Description

Product #

Add to Cart

Benzene SDS    
tert-Butyl methyl ether SDS    
Ethylbenzene SDS    
Toluene SDS    
o-Xylene SDS    
m-Xylene SDS    
p-Xylene SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1230 3/PG 2
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

CRM47505 BTEX/MTBE Mix, HC, certified reference material, TraceCERT®, 2000 μg/mL each component in methanol

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?