EC Number 216-653-1
| Related Categories | 500 Series Drinking Water Methods, Analytical Standards, Analytical/Chromatography, EPA, Method 552, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| feature | standard type surrogate |
| packaging | ampule of 1 mL |
| concentration | 1000 μg/mL in methyl tert-butyl ether |
| suitability | suitable for 552.2 per US EPA |
| storage temp. | 2-8°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?