EC Number 200-838-9
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Applications, Biochemicals and Reagents, |
| grade | analytical standard |
| packaging | pkg of 1 mL |
| concentration | 10 mg/mL in methylene chloride (as total weight) |
| storage temp. | −20°C |
Live Chat and Frequently Asked Questions
are available for this Product.
Qualitative mixture. Useful for establishing a retention time identification. Data sheet supplied with each ampule.
|
Description |
Product # |
Add to Cart |
|---|---|---|
| cis-9,cis-12-Octadecadienoic acid methyl ester ~ 10 % (w/w) | ||
| cis-9,trans-12-Octadecadienoic acid methyl ester ~ 20 % (w/w) | ||
| trans-9,cis-12-Octadecadienoic acid methyl ester ~ 20 % (w/w) | ||
| trans-9, trans-12-Octadecadienoic methyl ester ~ 50 % (w/w) |
Customers Also Viewed
10 mg/mL in methylene chloride (as total weight), analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?