Synonym: Linolenic Acid Methyl Ester solution
CAS Number 301-00-8
| Related Categories | Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Applications, Biochemicals and Reagents, |
| grade | analytical standard |
| packaging | ampule of 1 mL |
| concentration | 10 mg/mL in methylene chloride (as total weight) |
| storage temp. | −20°C |
Qualitative mixtures. Useful for establishing a retention time identification. Data sheet supplied with each ampule.
|
Description |
Product # |
Add to Cart |
|---|---|---|
| cis-9,cis-12,cis-15-Octadecatrienoic acid methyl ester ~ 3 % (w/w) | ||
| cis-9,cis-12,trans-15-Octadecatrienoic acid methyl ester ~ 7 % (w/w) | ||
| cis-9,trans-12,cis-15-Octadecatrienoic acid methyl ester ~ 7 % (w/w) | ||
| cis-9,trans-12,trans-15-Octadecatrienoic acid methyl ester ~ 15 % (w/w) | ||
| trans-9,cis-12,cis-15-Octadecatrienoic acid methyl ester ~ 7 % (w/w) | ||
| trans-9,cis-12,trans-15-Octadecatrienoic acid methyl ester ~ 15 % (w/w) | ||
| trans-9,trans-12,cis-15-Octadecatrienoic acid methyl ester ~ 15 % (w/w) | ||
| trans-9,trans-12,trans-15-Octadecatrienoic acid methyl ester ~ 30 % (w/w) |
Customers Also Viewed
10 mg/mL in methylene chloride (as total weight), analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?