CAS Number 460-00-4
Empirical Formula (Hill Notation) C6H4BrF
Molecular Weight 175.00
EC Number 200-659-6
| Related Categories | A-B, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 1000 μg/mL in methanol |
| storage temp. | 2-8°C |
Customers Also Viewed
2000 μg/mL in methanol, analytical standard
2000 μg/mL each component in methanol, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?