• USA Home
  • 47960-U - EPA 8270 Surrogate Standard

EMAIL THIS PAGE TO A FRIEND
47960-U Supelco

EPA 8270 Surrogate Standard

4000 μg/mL each component in methylene chloride, analytical standard

Synonym: EPA 8270 Surrogate Standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, E, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type surrogate
packaging   ampule of 1 mL
concentration   4000 μg/mL each component in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8270 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This product is now available as a certified reference solution. Please reference CRM47960.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

2-Fluorobiphenyl SDS    
2-Fluorophenol SDS    
Nitrobenzene-d5 SDS    
Phenol-d6 SDS    
p-Terphenyl-d14 SDS    
2,4,6-Tribromophenol SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM47960 EPA 8270 Surrogate Standard, certified reference material, TraceCERT®, 4000 μg/mL each component in dichloromethane

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?