• USA Home
  • 47964 - Aroclor 1221 solution

EMAIL THIS PAGE TO A FRIEND
47964 Supelco

Aroclor 1221 solution

500 mg/kg in transformer oil, analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aroclors, Aroclors, PCBs, and Dioxins,
grade   analytical standard
packaging   ampule of 5 mL
concentration   500 mg/kg in transformer oil
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
294.8 °F
Flash Point(C) 
146 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?