• USA Home
  • 47986 - MCPP methyl ester solution

EMAIL THIS PAGE TO A FRIEND
47986 Supelco

MCPP methyl ester solution

2000 μg/mL in hexane, analytical standard

Purchase

Properties

Related Categories 8000 Series Solidwaste Methods, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL in hexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8150 per US EPA
  suitable for 8151 per US EPA
  suitable for 8151A per US EPA
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1208 - class 3 - PG 2 - Hexanes
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Agricultural Pesticides on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of Agricultural Pesticides on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Metabolites, Pesticides, Purification, Size-exclusion chromatography

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?