CAS Number 12672-29-6
| Related Categories | 2000/60/EC, A, AR to AZ, Alphabetical Index of Analytical Standards, Analytical Standards, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 1000 μg/mL in methanol |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?