• USA Home
  • 48131 - MISA Group 20 Phenols Mix B

EMAIL THIS PAGE TO A FRIEND
48131 Supelco

MISA Group 20 Phenols Mix B

certified reference material, 2000 μg/mL each component in methanol

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments, Canada,
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for Group 20 per MISA (Canada)
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Analyte

Description

Product #

Add to Cart

4-Chloro-3-methylphenol SDS    
2-Chlorophenol SDS    
m-Cresol SDS    
2,4-Dichlorophenol SDS    
2,4-Dinitrophenol SDS    
4-Nitrophenol SDS    
Pentachlorophenol SDS    
Phenol SDS    
2,3,4,5-Tetrachlorophenol SDS    
2,4,6-Trichlorophenol SDS    
Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Phenolic Calibration Standards for Monitoring Wastewaters

Phenolic compounds are important raw materials for the manufacture of plastic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints and general disinfectants. Because of the toxicity ...
Vicki Yearick
AnalytiX Volume 6 Article 3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations, Size-exclusion chromatography

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?