• USA Home
  • 48137 - trans-Nonachlor solution

EMAIL THIS PAGE TO A FRIEND
48137 Supelco

trans-Nonachlor solution

100 μg/mL in hexane, analytical standard

  • CAS Number 39765-80-5

  • Empirical Formula (Hill Notation) C10H5Cl9

  • Molecular Weight 444.22

  •  Beilstein Registry Number 8158568

  •  EC Number 203-777-6

  •  MDL number MFCD00152673

  •  PubChem Substance ID 329757354

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   OCHOKXCPKDPNQU-FHPNKJTRSA-N
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   100 μg/mL in hexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN1208 - class 3 - PG 2 - Hexanes, solution
WGK Germany 
2
Flash Point(F) 
-14.8 °F
Flash Point(C) 
-26 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000

From our library of Articles, Sigma-Aldrich presents US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO™-1000
Keywords: Air Monitoring, Chromatography, Electron capture detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?