• USA Home
  • 48240-U - EPA 607 Nitrosamines Mix

EMAIL THIS PAGE TO A FRIEND
48240-U Supelco

EPA 607 Nitrosamines Mix

certified reference material, 2000 μg/mL each component in methanol

Purchase

Properties

Related Categories 600 Series Wastewater Methods, 8000 Series Solidwaste Methods, Additional Standards, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 607 per US EPA
  suitable for 8070 per US EPA
  suitable for 8270 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This certified reference material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Analyte

Description

Product #

Add to Cart

N-Nitrosodimethylamine SDS    
N-Nitrosodiphenylamine SDS    
N-Nitrosodi-n-propylamine SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?