CAS Number 15972-60-8
Empirical Formula (Hill Notation) C14H20ClNO2
Molecular Weight 269.77
Beilstein Registry Number 2944476
MDL number MFCD00041817
PubChem Substance ID 24871866
| Related Categories | 2000/60/EC, A, A-B, AA to AL, All alphabetically sorted, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 200 μg/mL in methanol |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?