48344 Supelco

Bromochloromethane solution

200 μg/mL in methanol, analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 74-97-5

  • Empirical Formula (Hill Notation) CH2BrCl

  • Molecular Weight 129.38

  •  EC Number 200-659-6

Properties

Related Categories A-B, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
packaging   ampule of 1 mL
concentration   200 μg/mL in methanol
storage temp.   2-8°C

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

Bromochloromethane

contains 50 ppm BHT as inhibitor (may contain up to), ≥99.5%

Bromochloromethane

analytical standard

2000 μg/mL in methanol, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?