• USA Home
  • 48460 - Pesticide Surrogate Spike Mix

EMAIL THIS PAGE TO A FRIEND
48460 Supelco

Pesticide Surrogate Spike Mix

200 μg/mL each component in acetone, analytical standard

Purchase

Properties

grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type surrogate
packaging   ampule of 1 mL
concentration   200 μg/mL each component in acetone
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8081 per US EPA
format   multi-component solution

Description

Other Notes

This product is now available as a certified reference material. Please reference CRM48460.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Decachlorobiphenyl SDS    
2,4,5,6-Tetrachloro-m-xylene SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1090 - class 3 - PG 2 - Acetone, solution
WGK Germany 
1
Flash Point(F) 
1.4 °F
Flash Point(C) 
-17 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM48460 Pesticide Surrogate Spike Mix, certified reference material, TraceCERT®, 200 μg/mL each component in acetone, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?