• USA Home
  • 48467 - EPA 8270 Benzidines Mix

EMAIL THIS PAGE TO A FRIEND
48467 Supelco

EPA 8270 Benzidines Mix

2000 μg/mL each component in methanol, analytical standard

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, E, Organic Pollutant Standards, Volatile & Semivolatile Standards More...
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 8270 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This product is now available as a certified reference material. Please reference CRM48467.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Benzidine SDS    
3,3′-Dichlorobenzidine SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

CRM48467 EPA 8270 Benzidines Mix, certified reference material, TraceCERT®, 2000 μg/mL each component in methanol, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?