48526 Supelco

1,1-Dichloroethene

analytical standard

DOWNLOAD MSDS (PDF)

Synonym: 1,1-Dichloroethylene, Vinylidene chloride

  • CAS Number 75-35-4

  • Linear Formula CH2=CCl2

  • Molecular Weight 96.94

  •  Beilstein Registry Number 1733365

  •  EC Number 200-864-0

  •  MDL number MFCD00011653

  •  PubChem Substance ID 24872048

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
vapor density   3.46 (vs air)
vapor pressure   9.68 psi ( 20 °C)
autoignition temp.   968 °F
expl. lim.   15.5 %
packaging   ampule of 5000 mg
bp   30-32 °C(lit.)
mp   −122 °C(lit.)
density   1.213 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

1,1-Dichloroethane

analytical standard

1000 μg/mL in methanol, analytical standard

1,1-Dichloroethylene

analytical standard

1,1-Dichloroethylene

contains 200 ppm MEHQ as inhibitor, 99%

<I>cis</I>-1,2-Dichloroethene

analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?