EMAIL THIS PAGE TO A FRIEND
48528 Supelco

2,4-Dichlorophenol

analytical standard

  • CAS Number 120-83-2

  • Linear Formula Cl2C6H3OH

  • Molecular Weight 163.00

  •  Beilstein Registry Number 742467

  •  EC Number 204-429-6

  •  MDL number MFCD00002169

  •  PubChem Substance ID 24872052

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   HFZWRUODUSTPEG-UHFFFAOYSA-N
form   neat
packaging   ampule of 5000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   209-210 °C(lit.)
mp   42-43 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2928 - class 6.1 - PG 2 - EHS - RQ - acidic - Toxic solids, corrosive, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
SK8575000
Flash Point(F) 
237.2 °F
Flash Point(C) 
114 °C
Protocols & Articles

Articles

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI-Chrom P
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Sample preparations, Size-exclusion chromatography, Solid phase extractions

GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols in Water on a 5% Phenyl Column after SPME using 85 μm Polyacrylate Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phenols on SLB®-5ms after SPME using 60 μm Carbowax® Fiber
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography, Solid phase microextractions

HPLC Analysis of Phenols on SUPELCOSIL™ LC-8

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Phenols on SUPELCOSIL™ LC-8
Keywords: Chromatography, High performance liquid chromatography

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations

US EPA Method 8041: GC Analysis of Phenols on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8041: GC Analysis of Phenols on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?