EMAIL THIS PAGE TO A FRIEND
48547 Supelco

Nitrobenzene

analytical standard

  • CAS Number 98-95-3

  • Empirical Formula (Hill Notation) C6H5NO2

  • Molecular Weight 123.11

  •  Beilstein Registry Number 507540

  •  EC Number 202-716-0

  •  MDL number MFCD00007043

  •  PubChem Substance ID 24872079

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
vapor density   4.2 (vs air)
vapor pressure   0.15 mmHg ( 20 °C)
  50 mmHg ( 120 °C)
InChI Key   LQNUZADURLCDLV-UHFFFAOYSA-N
form   neat
autoignition temp.   899 °F
expl. lim.   40 %
packaging   ampule of 1000 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.551(lit.)
bp   210-211 °C(lit.)
mp   5-6 °C(lit.)
density   1.196 g/mL at 25 °C(lit.)
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Target organs 
Blood
RIDADR 
UN 1662 6.1 / PGII
WGK Germany 
3
RTECS 
DA6475000
Flash Point(F) 
190.4 °F
Flash Point(C) 
88 °C
Protocols & Articles

Articles

EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18

From our library of Articles, Sigma-Aldrich presents EPA Method 8330: HPLC Analysis of Explosives on Ascentis® Express C18
Keywords: Chromatography, High performance liquid chromatography

GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Explosives in Water on a 14% Cyanopropylphenyl Column after SPME using 65 μm PDMS/DVB Fiber
Keywords: Chromatography, Electron capture detector, Gas chromatography, Purification, Solid phase microextractions

HPLC Analysis of Nitroaromatic Compounds Using Ascentis® Express Biphenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Nitroaromatic Compounds Using Ascentis® Express Biphenyl
Keywords: Chromatography, High performance liquid chromatography, Separation

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18

From our library of Articles, Sigma-Aldrich presents US EPA Method 8330: HPLC Analysis of Nitroaromatics and Nitramines, Mix A on Ascentis® C18
Keywords: Chromatography, Environmental, Environmental Analysis, High performance liquid chromatography

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Reliable Identification of F&F Components by GC/MS and Linear Retention Indices and Multidimensional Chromatography [VIDEO]

In this on-demand webinar by Prof. Luigi Mondello of the University of Messina, Italy, highlights the shortcomings of GC/MS analysis in the identification of F&F compounds when using a straightforwar...
Keywords: Chromatography, Gas chromatography, High performance liquid chromatography, Mass spectrometry

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?