EMAIL THIS PAGE TO A FRIEND
48588 Supelco

Aroclor 1232

analytical standard, for environmental analysis

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aroclors, Aroclors, PCBs, and Dioxins,
grade   analytical standard, for environmental analysis
form   neat
packaging   pkg of 10 mg
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2315 9 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?