48607 Supelco

1,1,2-Trichloroethane solution

200 μg/mL in methanol, analytical standard

DOWNLOAD MSDS (PDF)
  • CAS Number 79-00-5

  •  EC Number 200-659-6

Properties

Related Categories Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, Chemical Class,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   200 μg/mL in methanol

Price and Availability

Proficiency Testing Samples from RTC


PT Samples from RTCRTC has been producing environmental LPTP for more than 20 years and send out over 20,000 PT samples a year, throughout
the world.





Customers Also Viewed

1,1,2-<WBR>Trichloroethane

contains ≥3% 2-propanol as stabilizer, 97%

5000 μg/mL in methanol, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?