Synonym: Ethyl chloride
CAS Number 75-00-3
Empirical Formula (Hill Notation) C2H5Cl
Molecular Weight 64.51
EC Number 200-659-6
MDL number MFCD00000961
PubChem Substance ID 24871981
| Related Categories | Allergens, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 200 μg/mL in methanol |
| storage temp. | 2-8°C |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?