CAS Number 83-32-9
Beilstein Registry Number 386081
EC Number 200-659-6
MDL number MFCD00003807
PubChem Substance ID 24872166
| Related Categories | A, A-B, AA to AL, All PAH Standards, Alpha Sort, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 200 μg/mL in methanol |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?