• USA Home
  • 48662 - Fluoranthene solution

EMAIL THIS PAGE TO A FRIEND
48662 Supelco

Fluoranthene solution

certified reference material, 200 μg/mL in methylene chloride

Purchase

Properties

Related Categories Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   certified reference material
InChI Key   GVEPBJHOBDJJJI-UHFFFAOYSA-N
grade   TraceCERT®
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of × 1 mL
concentration   200 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?