• USA Home
  • 48714-U - Pyridine-d5 solution

EMAIL THIS PAGE TO A FRIEND
48714-U Supelco

Pyridine-d5 solution

2000 μg/mL in methylene chloride, analytical standard

Synonym: Pentadeuteropyridine

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, EPA,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type internal/surrogate
packaging   ampule of 1 mL
concentration   2000 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 625 per US EPA
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?