• USA Home
  • 48718 - Pentafluorophenol solution

EMAIL THIS PAGE TO A FRIEND
48718 Supelco

Pentafluorophenol solution

2000 μg/mL in methylene chloride, analytical standard

Synonym: 2,3,4,5,6-Pentafluorophenol solution

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type internal/surrogate
packaging   ampule of 1 mL
concentration   2000 μg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 625 per US EPA
format   single component solution
storage temp.   room temp

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

Phenolic Calibration Standards

Phenolic compounds are important raw materials for the manufacturing of phenolic resins, pharmaceuticals, laboratory reagents, dyes, germicidal paints, and general disinfectants. Due to their toxicit...
Reporter US Volume 28.3
Keywords: Chromatography, Environmental, Gas chromatography, Pharmaceutical, Sample preparations

US EPA Method 8041: GC Analysis of Phenols on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8041: GC Analysis of Phenols on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?