CAS Number 92-87-5
Empirical Formula (Hill Notation) C12H12N2
Molecular Weight 184.24
| Related Categories | A-B, Alpha Sort, Alphabetical Index of Analytical Standards, Analytical Standards, Analytical/Chromatography, |
| grade | analytical standard |
| CofA | certificate of analysis is enclosed in each package. |
| packaging | ampule of 1 mL |
| concentration | 500 μg/mL in methylene chloride |
| storage temp. | 2-8°C |
Customers Also Viewed
5000 μg/mL in methanol, analytical standard
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?