• USA Home
  • 48736 - Aroclor 1260 solution

EMAIL THIS PAGE TO A FRIEND
48736 Supelco

Aroclor 1260 solution

50 mg/kg in transformer oil, analytical standard

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Analytical Standards, Analytical/Chromatography, Aroclors,
grade   analytical standard
packaging   ampule of 5 mL
concentration   50 mg/kg in transformer oil
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution

Description

Other Notes

This product is also available as a certified reference material (CRM). Please see product CRM48736 for details and ordering.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

CRM48736 Aroclor 1260, certified reference material, 50 mg/kg in transformer oil, pkg of 1 × 5 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?