• USA Home
  • 48743 - EPA 610 Polynuclear Aromatic Hydrocarbons Mixture

EMAIL THIS PAGE TO A FRIEND
48743 Supelco

EPA 610 Polynuclear Aromatic Hydrocarbons Mixture

analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Column Test Mixes (HPLC),
grade   analytical standard
description   LC-PAH
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   in methanol: methylene chloride (1:1)
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 5506 per NIOSH
  suitable for 5515 per NIOSH
  suitable for 610 per US EPA
  suitable for IP-7 per US EPA - TO
  suitable for TO-13 per EPA - TO
compatibility   use to QA LC-PAH columns
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


All labs need water

Analyte

Description

Product #

Add to Cart

Acenaphthene 1000 μg/mL SDS    
Acenaphthylene 2000 μg/mL SDS    
Anthracene 100 μg/mL SDS    
Benz[a]anthracene 100 μg/mL SDS    
Benzo[b]fluoranthene 200 μg/mL SDS    
Benzo[k]fluoranthene 100 μg/mL SDS    
Benzo[ghi]perylene 200 μg/mL SDS    
Benzo[a]pyrene 100 μg/mL SDS    
Chrysene 100 μg/mL SDS    
Dibenz[a,h]anthracene 200 μg/mL SDS    
Fluoranthene 200 μg/mL SDS    
Fluorene 200 μg/mL SDS    
Indeno[1,2,3-cd]pyrene 100 μg/mL SDS    
Naphthalene 1000 μg/mL SDS    
Phenanthrene 100 μg/mL SDS    
Pyrene 100 μg/mL SDS    
Safety & Documentation

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Supelco HPLC Calculator for Isocratic or Gradient Method Transfer

Calculates conditions for transfer of an isocratic or gradient method from one HPLC column to another. Allows method scaling from microbore through preparative column range. Calculates time and solve...
Keywords: High performance liquid chromatography

Related Products

replaced by

Product #

Description

Add to Cart

CRM48743 EPA 610 Polynuclear Aromatic Hydrocarbons Mix, certified reference material, in methanol: methylene chloride (1:1) (varied concentration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?