EMAIL THIS PAGE TO A FRIEND
48777 Supelco

EPA VOC Mix 2

2000 μg/mL each component in methanol, analytical standard

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography, Drinking Water,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
packaging   ampule of 1 mL
concentration   2000 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 502.1 per US EPA
  suitable for 502.2 per US EPA
  suitable for 503.1 per US EPA
  suitable for 524.1 per US EPA
  suitable for 524.2 per US EPA
  suitable for 8021 (1987) per US EPA
  suitable for 8021A (1990) per US EPA
  suitable for 8260 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Footnote

Separate Source standard. Prepared from independently sourced raw material and quality controlled independently.

Price and Availability

Analyte

Description

Product #

Add to Cart

Benzene SDS    
Bromobenzene SDS    
Butylbenzene SDS    
Ethylbenzene SDS    
Naphthalene SDS    
Styrene SDS    
Toluene SDS    
1,2,3-Trichlorobenzene SDS    
1,2,4-Trichlorobenzene SDS    
1,2,4-Trimethylbenzene SDS    
m-Xylene SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

replaced by

Product #

Description

Add to Cart

CRM48777 EPA VOC Mix 2, certified reference material, 2000 μg/mL each component in methanol, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?