• USA Home
  • 48831 - EPA 625 Base-Neutral 1

EMAIL THIS PAGE TO A FRIEND
48831 Supelco

EPA 625 Base-Neutral 1

certified reference material, 200 μg/mL each component in methylene chloride

Synonym: EPA 625 Base/Neutral 1 in Methylene Chloride

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Application Areas and Market Segments,
grade   certified reference material
  TraceCERT®
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   200 μg/mL each component in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 625 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO Guide 34:2009 and ISO/IEC 17025:2005. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Analyte

Description

Product #

Add to Cart

Acenaphthylene SDS    
Benzo[b]fluoranthene SDS    
Bis(2-chloroethyl) ether SDS    
Bis(2-ethylhexyl) phthalate SDS    
4-Bromodiphenyl ether SDS    
Bis-(2-chloroisopropyl) ether SDS    
Dibutyl phthalate SDS    
1,4-Dichlorobenzene SDS    
3,3′-Dichlorobenzidine SDS    
Dimethyl phthalate SDS    
2,6-Dinitrotoluene SDS    
Nitrobenzene SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Precautionary statements 
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?