EMAIL THIS PAGE TO A FRIEND
48851-U Supelco

EPA Purgeable A

analytical standard, 200 μg/mL each component in methanol

Purchase

Properties

Related Categories 600 Series Wastewater Methods, Additional Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
description   Separate Source
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 × 1 mL
concentration   200 μg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for 624 per US EPA
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Carbon tetrachloride SDS    
Chlorobenzene SDS    
2-Chloroethyl vinyl ether SDS    
Chloroform SDS    
Dibromochloromethane SDS    
1,1-Dichloroethane SDS    
1,1-Dichloroethylene SDS    
Dichloromethane SDS    
1,2-Dichloropropane SDS    
Tetrachloroethylene SDS    
1,1,2-Trichloroethane SDS    
Trichloroethylene SDS    
Trichlorofluoromethane SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?