• USA Home
  • 48891 - Ethylene oxide solution

EMAIL THIS PAGE TO A FRIEND
48891 Supelco

Ethylene oxide solution

50 mg/mL in methylene chloride, analytical standard

  • CAS Number 75-21-8

  • Empirical Formula (Hill Notation) C2H4O

  • Molecular Weight 44.05

  •  MDL number MFCD00014482

  •  PubChem Substance ID 329757420

Purchase

Properties

Related Categories Additional Standards, Air Monitoring Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   IAYPIBMASNFSPL-UHFFFAOYSA-N
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   50 mg/mL in methylene chloride
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
Target organs 
Blood, Central nervous system, Liver
RIDADR 
UN 1593 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

replaced by

Product #

Description

Add to Cart

CRM48891 Ethylene oxide solution, certified reference material, 50 mg/mL in dichloromethane, ampule of 1 mL

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?