• USA Home
  • 48897 - EPA TO-1 Toxic Organic Mix 1B

EMAIL THIS PAGE TO A FRIEND
48897 Supelco

EPA TO-1 Toxic Organic Mix 1B

2 mg/mL each component in methanol, analytical standard

Purchase

Properties

Related Categories Additional Standards, Air Monitoring Standards, Analytical Standards, Analytical/Chromatography, Chromatography,
grade   analytical standard
CofA   certificate of analysis is enclosed in each package.
feature   standard type calibration
packaging   ampule of 1 mL
concentration   2 mg/mL each component in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
suitability   suitable for IP-1 per US EPA - TO
  suitable for TO-1 per EPA - TO
  suitable for TO-2 per EPA - TO
  suitable for TO-3 per EPA - TO
format   multi-component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Analyte

Description

Product #

Add to Cart

Acrylonitrile SDS    
Allyl chloride SDS    
Bromobenzene SDS    
Bromoform SDS    
Carbon tetrachloride SDS    
Chlorobenzene SDS    
Chloroform SDS    
1,2-Dibromoethane SDS    
1,2-Dichloroethane SDS    
1,2-Dichloropropane SDS    
1,3-Dichloropropane SDS    
Tetrachloroethylene SDS    
1,1,1-Trichloroethane SDS    
Trichloroethylene SDS    
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
3
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?